(E)-4-(4-Fluorophenyl)-4-oxobut-2-enoic acid 97%

Supplier: Ambeed
Warning

A423222-100MG A423222-1G A423222-250MG
77252-352EA 27.81 USD
77252-352 77252-354 77252-356
(E)-4-(4-Fluorophenyl)-4-oxobut-2-enoic acid 97%
(E)-4-(4-Fluorophenyl)-4-oxobut-2-enoic acid

Precaución: for Research Use Only

Formula: C₁₀H₇FO₃
MW: 194.15 g/mol
MDL Number: MFCD01656421
CAS Number: 35504-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR