2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-7-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A423126-100MG
A423126-1G
A423126-250MG
A423126-5G
77252-344EA
12
USD
77252-344
77252-346
77252-348
77252-350
2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-7-carboxylic acid 98%
2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-7-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₄ |
MDL Number:
MFCD03094736 CAS Number: 149353-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...