2-Chloro-quinazolin-8-ol 98%

Supplier: Ambeed
Danger

A422904-100MG A422904-1G A422904-250MG
77252-332EA 19 USD
77252-332 77252-334 77252-336
2-Chloro-quinazolin-8-ol 98%
2-Chloro-quinazolin-8-ol

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MDL Number: MFCD24728919
CAS Number: 953039-10-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR