Mono-O-AcetylGanciclovir 97%

Supplier: Ambeed
Warning

A422387-100MG A422387-1G A422387-5G
77252-326EA 5.15 USD
77252-326 77252-328 77252-330
Mono-O-AcetylGanciclovir 97%
Mono-O-AcetylGanciclovir

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₅O₅
MW: 297.26 g/mol
MDL Number: MFCD09840659
CAS Number: 88110-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR