(14aR)-1,14-Bis(diphenylphosphino)-6,7,8,9-tetrahydrodibenzo[b,d][1,6]dioxecine 98%

Supplier: Ambeed
Warning

A422300-1G A422300-250MG
77264-740EA 713.99 USD
77264-740 77264-742
(14aR)-1,14-Bis(diphenylphosphino)-6,7,8,9-tetrahydrodibenzo[b,d][1,6]dioxecine 98%
(14aR)-1,14-Bis(diphenylphosphino)-6,7,8,9-tetrahydrodibenzo[b,d][1,6]dioxecine

Precaución: for Research Use Only

Formula: C₄₀H₃₄O₂P₂
CAS Number: 301847-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR