1-(2,2,2-Trifluoroethyl)-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(2,2,2-trifluoroethyl)-3,6-dihydro-2H-pyridine, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(2,2,2-trifluoroethyl)-1,2,3,6-tetrahydropyridine

A422264-100MG A422264-1G A422264-250MG
77264-734EA 41.2 USD
77264-734 77264-736 77264-738
1-(2,2,2-Trifluoroethyl)-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester 98%
1-(2,2,2-Trifluoroethyl)-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₁BF₃NO₂
MW: 291.118 g/mol
MDL Number: MFCD18383322
CAS Number: 1219931-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR