6-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-c]pyridin-7(6H)-one 95%

Supplier: Ambeed
Warning

A421945-100MG A421945-1G A421945-250MG A421945-50MG
77264-714EA 157.59 USD
77264-714 77264-716 77264-718 77264-720
6-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-c]pyridin-7(6H)-one 95%
6-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-c]pyridin-7(6H)-one

Precaución: for Research Use Only

Formula: C₂₁H₂₅BN₂O₅S
MW: 428.30 g/mol
MDL Number: MFCD30802581
CAS Number: 1445993-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR