(4R,4'R)-2,2'-(Pentane-3,3-diyl)bis(4-phenyl-4,5-dihydrooxazole) 98%

Supplier: Ambeed
Warning

A421892-100MG A421892-1G A421892-250MG A421892-50MG
77264-706EA 53.56 USD
77264-706 77264-708 77264-710 77264-712
(4R,4'R)-2,2'-(Pentane-3,3-diyl)bis(4-phenyl-4,5-dihydrooxazole) 98%
(4R,4'R)-2,2'-(Pentane-3,3-diyl)bis(4-phenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₂
MW: 362.46 g/mol
MDL Number: MFCD31010439
CAS Number: 195515-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR