PF-3084014 99+%

Supplier: Ambeed
Warning

Synonyms: Nirogacestat, (S)-2-(((S)-6,8-Difluoro-1,2,3,4-tetrahydronaphthalen-2-yl)amino)-N-(1-(2-methyl-1-(neopentylamino)propan-2-yl)-1H-imidazol-4-yl)pentanamide

A421613-10MG A421613-1MG A421613-25MG A421613-5MG
77240-822EA 77 USD
77240-822 77240-824 77240-826 77240-828
PF-3084014 99+%
PF-3084014

Precaución: for Research Use Only

Formula: C₂₇H₄₁F₂N₅O
MW: 489.60 g/mol
MDL Number: MFCD19980705
CAS Number: 1290543-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR