2-(3-(Trifluoromethyl)phenyl)ethanol 97%

Supplier: Ambeed
Warning

A421517-1G A421517-250MG A421517-5G
77278-966EA 33 USD
77278-966 77278-968 77278-970
2-(3-(Trifluoromethyl)phenyl)ethanol 97%
2-(3-(Trifluoromethyl)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
MDL Number: MFCD00009710
CAS Number: 455-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR