PF-06409577 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-5-(4-(1-hydroxycyclobutyl)phenyl)-1H-indole-3-carboxylic acid, 6-Chloro-5-[4-(1-hydroxycyclobutyl)phenyl]-1H-indole-3-carboxylic acid

A421471-10MG A421471-1MG A421471-25MG A421471-50MG A421471-5MG
77264-696EA 52 USD
77264-696 77264-698 77264-700 77264-702 77264-704
PF-06409577 97%
PF-06409577

Precaución: for Research Use Only

Formula: C₁₉H₁₆ClNO₃
MW: 341.79 g/mol
MDL Number: MFCD29917835
CAS Number: 1467057-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR