(4-Bromophenyl)acetone 98%

Supplier: Ambeed
Warning

A421422-100G A421422-1G A421422-5G
77291-418EA 430.01 USD
77291-418 77291-420 77291-422
(4-Bromophenyl)acetone 98%
(4-Bromophenyl)acetone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD00210401
CAS Number: 6186-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR