2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine 95%

Supplier: Ambeed
Warning

A421216-100MG A421216-1G A421216-250MG
77240-816EA 24 USD
77240-816 77240-818 77240-820
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine 95%
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₀ClN₃
MDL Number: MFCD02255631
CAS Number: 135875-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR