3-(3,5-Dichlorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A420391-100G A420391-100MG A420391-1G A420391-250MG A420391-25G A420391-5G
77278-954EA 478 USD
77278-954 77278-956 77278-958 77278-960 77278-962 77278-964
3-(3,5-Dichlorophenyl)propionic acid 97%
3-(3,5-Dichlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MDL Number: MFCD04037332
CAS Number: 95333-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR