1-Iodo-4-(trans-4-pentylcyclohexyl)benzene 95%

Supplier: Ambeed
Warning

A420291-1G A420291-25G A420291-5G
77291-406EA 10.3 USD
77291-406 77291-408 77291-410
1-Iodo-4-(trans-4-pentylcyclohexyl)benzene 95%
1-Iodo-4-(trans-4-pentylcyclohexyl)benzene

Precaución: for Research Use Only

Formula: C₁₇H₂₅I
MW: 356.28 g/mol
MDL Number: MFCD06658190
CAS Number: 116963-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR