TCS 401 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Carboxyformamido)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid hydrochloride

A420041-100MG A420041-10MG A420041-1MG A420041-25MG A420041-50MG A420041-5MG
77264-682EA 413.03 USD
77264-682 77264-684 77264-686 77264-688 77264-690 77264-692
TCS 401 98%
TCS 401

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂O₅S
MW: 306.72 g/mol
MDL Number: MFCD09971132
CAS Number: 243966-09-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR