4-(1-Naphthyl)-3,5-diphenyl-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A419635-100MG A419635-1G A419635-250MG
77278-948EA 21 USD
77278-948 77278-950 77278-952
4-(1-Naphthyl)-3,5-diphenyl-1,2,4-triazole 98%
4-(1-Naphthyl)-3,5-diphenyl-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₂₄H₁₇N₃
MDL Number: MFCD09832971
CAS Number: 16152-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR