((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: 5'-Tosyladenosine

A419540-250MG A419540-25G A419540-1G
77291-396EA 8.24 USD
77291-396 77291-398 77616-820
((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl 4-methylbenzenesulfonate 95%
((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₅O₆S
MW: 421.42 g/mol
MDL Number: MFCD00005751
CAS Number: 5135-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR