4-Fluoro-2-methylbenzylamine 97%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-2-methylphenyl)methanamine

A419520-100MG A419520-1G A419520-250MG A419520-25G A419520-5G
77291-386EA 7.21 USD
77291-386 77291-388 77291-390 77291-392 77291-394
4-Fluoro-2-methylbenzylamine 97%
4-Fluoro-2-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD03701060
CAS Number: 771574-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR