8-Bromo-11H-benzo[a]carbazole 97%

Supplier: Ambeed
Warning

A419426-250MG A419426-5G
77264-678EA 8 USD
77264-678 77264-680
8-Bromo-11H-benzo[a]carbazole 97%
8-Bromo-11H-benzo[a]carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₀BrN
MDL Number: MFCD27644691
CAS Number: 21064-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR