Ethyl (4-methoxybenzyl)carbamate 97%

Supplier: Ambeed
Warning

A419233-1G A419233-250MG A419233-5G
77240-810EA 22 USD
77240-810 77240-812 77240-814
Ethyl (4-methoxybenzyl)carbamate 97%
Ethyl (4-methoxybenzyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MDL Number: MFCD03392918
CAS Number: 35573-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR