21-Bromohenicosanoic acid 98+%

Supplier: Ambeed
Warning

A418813-100MG A418813-1G A418813-250MG
77252-314EA 25 USD
77252-314 77252-316 77252-318
21-Bromohenicosanoic acid 98+%
21-Bromohenicosanoic acid

Precaución: for Research Use Only

Formula: C₂₁H₄₁BrO₂
MDL Number: MFCD00082607
CAS Number: 855953-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR