4-(Chloromethyl)-7-hydroxy-2H-chromen-2-one 97%

Supplier: Ambeed
Danger

A417906-1G A417906-250MG A417906-5G
77291-372EA 60 USD
77291-372 77291-374 77748-846
4-(Chloromethyl)-7-hydroxy-2H-chromen-2-one 97%
4-(Chloromethyl)-7-hydroxy-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₀H₇ClO₃
MDL Number: MFCD00467168
CAS Number: 25392-41-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR