2-(Naphthalen-1-yl)ethanamine 95%

Supplier: Ambeed
Warning

A417399-100MG A417399-250MG A417399-5G
77264-670EA 8 USD
77264-670 77264-672 77551-258
2-(Naphthalen-1-yl)ethanamine 95%
2-(Naphthalen-1-yl)ethanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MDL Number: MFCD01529866
CAS Number: 4735-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR