GSK1904529A 98+%

Supplier: Ambeed
Warning

Synonyms: N-(2,6-Difluorophenyl)-5-(3-(2-((5-ethyl-2-methoxy-4-(4-(4-(methylsulphonyl)piperazin-1-yl)piperidin-1-yl)phenyl)amino)pyrimidin-4-yl)imidazo[1,2-a]pyridin-2-yl)-2-methoxybenzamide

A417175-1MG A417175-5MG
77240-798EA 77.25 USD
77240-798 77240-800
GSK1904529A 98+%
GSK1904529A

Precaución: for Research Use Only

Formula: C₄₄H₄₇F₂N₉O₅S
MW: 851.96 g/mol
MDL Number: MFCD17010271
CAS Number: 1089283-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR