3-Benzylrhodanine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyl-2-thioxo-4-thiazolidinone, 3-Benzyl-2-thioxothiazolidin-4-one

A417035-1G A417035-25G A417035-5G
77278-932EA 15.45 USD
77278-932 77278-934 77278-936
3-Benzylrhodanine 98%
3-Benzylrhodanine

Precaución: for Research Use Only

Formula: C₁₀H₉NOS₂
MW: 223.31 g/mol
MDL Number: MFCD00086924
CAS Number: 10574-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR