Isobutyldimethoxy(methyl)silane 95%

Supplier: Ambeed
Danger

A416438-10G A416438-25G A416438-5G
77252-280EA 36.05 USD
77252-280 77252-282 77252-284
Isobutyldimethoxy(methyl)silane 95%
Isobutyldimethoxy(methyl)silane

Precaución: for Research Use Only

Formula: C₇H₁₈O₂Si
MW: 162.30 g/mol
MDL Number: MFCD01320960
CAS Number: 18293-82-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR