2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole trihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Hoechst 33342 3HCl

A416033-1MG A416033-50MG A416033-10MG
77240-794EA 18.54 USD
77240-794 77240-796 77746-839
2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole trihydrochloride 98%
2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole trihydrochloride

Precaución: for Research Use Only

Formula: C₂₇H₃₁Cl₃N₆O
MW: 561.93 g/mol
MDL Number: MFCD30179380
CAS Number: 875756-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR