2-[(4R)-4,5-Dihydro-4-(phenylmethyl)-2-oxazolyl]pyridine 98%

Supplier: Ambeed
Warning

A415922-100MG A415922-1G A415922-250MG A415922-5G
77252-266EA 37.08 USD
77252-266 77252-268 77252-270 77252-272
2-[(4R)-4,5-Dihydro-4-(phenylmethyl)-2-oxazolyl]pyridine 98%
2-[(4R)-4,5-Dihydro-4-(phenylmethyl)-2-oxazolyl]pyridine

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O
MW: 238.28 g/mol
MDL Number: MFCD30541660
CAS Number: 1108603-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR