4-Fluorobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A415842-100G A415842-10G A415842-25G A415842-500G
77291-352EA 74 USD
77291-352 77291-354 77291-356 77748-841
4-Fluorobenzenesulfonamide 98%
4-Fluorobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₆FNO₂S
MDL Number: MFCD00025384
CAS Number: 402-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR