2-Bromo-1,3-bis(bromomethyl)benzene 95%

Supplier: Ambeed
Danger

A415170-1G A415170-250MG A415170-25G A415170-5G
77264-636EA 59 USD
77264-636 77264-638 77264-640 77264-642
2-Bromo-1,3-bis(bromomethyl)benzene 95%
2-Bromo-1,3-bis(bromomethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇Br₃
MDL Number: MFCD00229246
CAS Number: 25006-88-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR