4,4'-Bis(bromomethyl)biphenyl 95%

Supplier: Ambeed
Danger

A415093-1G A415093-5G
77291-348EA 24 USD
77291-348 77291-350
4,4'-Bis(bromomethyl)biphenyl 95%
4,4'-Bis(bromomethyl)biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂Br₂
MDL Number: MFCD00094419
CAS Number: 20248-86-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR