1-((2R,4R,5R)-3,3-Difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A415035-100MG A415035-1G A415035-250MG
77252-260EA 71.07 USD
77252-260 77252-262 77252-264
1-((2R,4R,5R)-3,3-Difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione 98%
1-((2R,4R,5R)-3,3-Difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₉H₁₀F₂N₂O₅
MW: 264.18 g/mol
MDL Number: MFCD00871912
CAS Number: 114248-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR