(4-Bromophenoxy)acetic acid 96%

Supplier: Ambeed
Warning

Synonyms: 4-Bromophenoxyacetic acid

A414744-100G A414744-10G A414744-25G A414744-500G
77240-776EA 54.59 USD
77240-776 77240-778 77240-780 77240-782
(4-Bromophenoxy)acetic acid 96%
(4-Bromophenoxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄Br)OCH₂COOH
MW: 231.05 g/mol
MDL Number: MFCD00093070
CAS Number: 1878-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR