2-Ethylhexyl methacrylate 99% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A414090-100G A414090-25G A414090-500G
77264-620EA 8 USD
77264-620 77264-622 77264-624
2-Ethylhexyl methacrylate 99% stabilized
2-Ethylhexyl methacrylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MDL Number: MFCD00009494
CAS Number: 688-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR