Sodium 4,4'-((3,5-dimethylphenyl)azanediyl)bis(butane-1-sulfonate) 98%

Supplier: Ambeed
Warning

A414039-100MG A414039-1G A414039-250MG
77291-332EA 37.08 USD
77291-332 77291-334 77291-336
Sodium 4,4'-((3,5-dimethylphenyl)azanediyl)bis(butane-1-sulfonate) 98%
Sodium 4,4'-((3,5-dimethylphenyl)azanediyl)bis(butane-1-sulfonate)

Precaución: for Research Use Only

Formula: C₁₆H₂₅NNa₂O₆S₂
MW: 437.48 g/mol
MDL Number: MFCD09038767
CAS Number: 209518-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR