2,3,4-Trifluoroaniline 98%

Supplier: Ambeed
Danger

A413761-100G A413761-10G A413761-25G A413761-5G
77291-324EA 20.6 USD
77291-324 77291-326 77291-328 77291-330
2,3,4-Trifluoroaniline 98%
2,3,4-Trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄F₃N
MW: 147.1 g/mol
MDL Number: MFCD00011737
CAS Number: 3862-73-5
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR