2-(6-Methoxypyridin-2-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 97%

Supplier: Ambeed
Warning

A413236-5G A413236-1G
77240-762EA 266.77 USD
77240-762 77746-837
2-(6-Methoxypyridin-2-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 97%
2-(6-Methoxypyridin-2-yl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₃BN₂O₅
MW: 264.04 g/mol
MDL Number: MFCD16621487
CAS Number: 1227700-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR