1-((3,5-Dimethyl-1H-pyrazol-1-yl)methyl)-1H-pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A413190-100MG A413190-1G A413190-250MG
77240-756EA 50.47 USD
77240-756 77240-758 77240-760
1-((3,5-Dimethyl-1H-pyrazol-1-yl)methyl)-1H-pyrazole-3-carboxylic acid 97%
1-((3,5-Dimethyl-1H-pyrazol-1-yl)methyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O₂
MW: 220.22 g/mol
MDL Number: MFCD04967757
CAS Number: 1004192-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR