Quipazine maleate 98%

Supplier: Ambeed
Danger

Synonyms: Quipazine dimaleate, 2-(Piperazin-1-yl)quinoline maleate

A413189-100MG A413189-1G A413189-250MG
77252-254EA 86 USD
77252-254 77252-256 77252-258
Quipazine maleate 98%
Quipazine maleate

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₄
MW: 329.356 g/mol
MDL Number: MFCD00133796
CAS Number: 5786-68-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR