4-Amino-3-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Danger

Synonyms: 2-Amino-5-cyanobenzotrifluoride

A413088-25G A413088-5G
77278-890EA 37.08 USD
77278-890 77278-892
4-Amino-3-(trifluoromethyl)benzonitrile 97%
4-Amino-3-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
MDL Number: MFCD00275473
CAS Number: 327-74-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR