3-Methylstyrene 97% stabilized

Supplier: Ambeed
Danger

Synonyms: 3-Vinyltoluene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A413069-5G A413069-1G
77291-322EA 78.28 USD
77291-322 77572-282
3-Methylstyrene 97% stabilized
3-Methylstyrene

Precaución: for Research Use Only

Formula: C₉H₁₀
MW: 118.18 g/mol
MDL Number: MFCD00008617
CAS Number: 100-80-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR