Tris(2-cyanoethyl)amine 99%

Supplier: Ambeed
Danger

A412706-1G A412706-250MG A412706-5G
77240-750EA 20 USD
77240-750 77240-752 77240-754
Tris(2-cyanoethyl)amine 99%
Tris(2-cyanoethyl)amine

Precaución: for Research Use Only

Formula: C₉H₁₂N₄
MDL Number: MFCD00671575
CAS Number: 7528-78-1
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR