4-Methyl-7-nitro-1H-indole 97%

Supplier: Ambeed
Warning

A412202-100MG A412202-1G
77291-316EA 62 USD
77291-316 77291-318
4-Methyl-7-nitro-1H-indole 97%
4-Methyl-7-nitro-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD07780760
CAS Number: 289483-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR