4-Isopropylcyclohexanol 98%

Supplier: Ambeed
Warning

A412175-100G A412175-25G A412175-5G
77291-310EA 219 USD
77291-310 77291-312 77291-314
4-Isopropylcyclohexanol 98%
4-Isopropylcyclohexanol

Precaución: for Research Use Only

Formula: C₉H₁₈O
MDL Number: MFCD00043476
CAS Number: 4621-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR