(E,E)-RGFP966 97%

Supplier: Ambeed
Warning

A411973-10MG A411973-1MG A411973-250MG A411973-25MG A411973-50MG A411973-5MG A411973-1G A411973-100MG
77264-596EA 44.01 USD
77264-596 77264-598 77264-600 77264-602 77264-604 77748-835 77748-834 77748-833
(E,E)-RGFP966 97%
(E,E)-RGFP966

Precaución: for Research Use Only

Formula: C₂₁H₁₉FN₄O
MDL Number: MFCD26960954
CAS Number: 1396841-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR