2,3-Diethylpyrazine 98+%

Supplier: Ambeed
Danger

A411703-1G A411703-25G A411703-5G
77252-232EA 10 USD
77252-232 77252-234 77252-236
2,3-Diethylpyrazine 98+%
2,3-Diethylpyrazine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MDL Number: MFCD00006151
CAS Number: 15707-24-1
UN: 3334
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR