1-(1-Butoxyethoxy)butane 95%

Supplier: Ambeed
Danger

A411647-250MG A411647-1G
77252-230EA 99 USD
77252-230 77594-258
1-(1-Butoxyethoxy)butane 95%
1-(1-Butoxyethoxy)butane

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₂
MDL Number: MFCD00129691
CAS Number: 871-22-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR