2,2-Bis(4-hydroxy-3-methylphenyl)propane 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(Propane-2,2-diyl)bis(2-methylphenol), 4,4'-Isopropylidenedi-o-cresol

A411546-100G A411546-25G A411546-500G A411546-5G
77252-222EA 108.15 USD
77252-222 77252-224 77252-226 77252-228
2,2-Bis(4-hydroxy-3-methylphenyl)propane 98%
2,2-Bis(4-hydroxy-3-methylphenyl)propane

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₂
MW: 256.35 g/mol
MDL Number: MFCD00002232
CAS Number: 79-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR