(3-Aminopropyl)triethoxysilane 98%

Supplier: Ambeed
Danger

Synonyms: 3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane

A411463-25G A411463-100G
77278-886EA 5.15 USD
77278-886 77601-792
(3-Aminopropyl)triethoxysilane 98%
(3-Aminopropyl)triethoxysilane

Precaución: for Research Use Only

Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
MDL Number: MFCD00008207
CAS Number: 919-30-2
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR